Eisco Clear Vinyl Aprons
: Wards
Heavy, 6.7 mil clear vinyl aprons protect clothing from stains and damage.
- Protects skin and clothing from lab reagents, acids, oils, and grease
- Made of heavy 6.7 mil vinyl - does not contain latex
- Halter design for unrestricted arm movement
- Reinforced at strain points to resist stretching
- Transparent
Suitable for labs or shop floors.
- Catalog No:
- 470345-690
- 470345-010
- Garment application:
- Office, Laboratory
- Office, Laboratory
- Gender:
- Unisex
- Unisex
- Fabric:
- Vinyl
- Vinyl
Frequently Bought Together