ContactUsLinkComponent
Drosophila Y-Maze
Drosophila Y-Maze
Catalog # 76670-414
Supplier:  MAZE ENGINEERS
undefined
Drosophila Y-Maze
Catalog # 76670-414
Supplier:  MAZE ENGINEERS
Supplier Number:  8502
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Drosophila Y-maze
  • Material:
    Acrylic
  • Environmentally Preferable:
  • Cat. No.:
    76670-414
Specifications
About this item
The Drosophila Y-Maze is a laboratory task used to study spatial learning and memory in Drosophila melanogaster.
  • Acrylic construction
  • Ten sets of 10 Y-Mazes
In this task, the fruit fly is placed at the end of one arm of a Y-shaped maze and is allowed to freely explore the maze. It is a simple and widely used experimental paradigm in which a fly is placed in a Y-shaped maze and given the opportunity to choose between two arms of the maze. The fly’s choice is recorded and the fly is then given a reward, such as food, in one of the arms. The fly is then placed back in the maze and given the opportunity to choose again.