ContactUsLinkComponent
Incline Rolling Ladder
Incline Rolling Ladder
Catalog # 76670-200
Supplier:  MAZE ENGINEERS
Incline Rolling Ladder
Catalog # 76670-200
Supplier:  MAZE ENGINEERS
Supplier Number:  5010
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Incline rolling ladder, mouse/rat
  • Material:
    Acrylic/metal
  • Environmentally Preferable:
  • Cat. No.:
    76670-200
Specifications
About this item
The incline rolling ladder test is a novel test sensitive to tactile and proprioception behaviors.
  • Acrylic frame and metal rods
  • 34 cm long ladder, with nine adjustable rungs
Developed to study functional deficits in rodents, the incline rolling ladder test consists of a ladder opening to a platform, set at an angle of 45 degrees with rungs that have an immobile textured part, and a movable rolling smooth part. The subjects are allowed to climb the ladder to reach the platform. The data is collected about the number of successful steps, the number of slips, and the specific parts (rungs) these occurred from.