About this item
Rabbit polyclonal antibody to Visinin Like Protein 1 for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.
- Validated Applications: WB, ICC/IF, IHC
- Recommended Dilutions: WB: 1:10000, ICC/IF: 1:2000, IHC: 1:200
Type: Primary
Antigen: Visinin Like Protein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77077-930
- 77080-276
- Antigen Symbol:
- Visinin Like Protein 1
- Visinin Like Protein 1
- Antigen Name:
- 21 kDa CABP
- 21 kDa CABP
- Antigen Synonyms:
- HPCAL 3|HPCAL3|Hippocalcin-like protein 3|Neural visinin-like type 1 protein|HLP3|Neural visinin-like protein 1|Hippocalcin like protein 3|HLP 3|HUVISL1
- HPCAL 3|HPCAL3|Hippocalcin-like protein 3|Neural visinin-like type 1 protein|HLP3|Neural visinin-like protein 1|Hippocalcin like protein 3|HLP 3|HUVISL1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# P62760
- UniprotID# P62760
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant full-length human VLP1, expressed in and purified from E. coli
- Recombinant full-length human VLP1, expressed in and purified from E. coli
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 18 kDa
- 18 kDa
- Purification:
- Immunogen affinity purification.
- Immunogen affinity purification.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested Applications:
- ICC/IF, IHC
- ICC/IF, IHC