About this item
Rabbit monoclonal [ARC0657] antibody to Glutamate Receptor 1 (AMPA subtype) for WB with samples derived from Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:200
Type: Primary
Antigen: Glutamate Receptor 1 (AMPA subtype)
Clonality: Monoclonal
Clone: ARC0657
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77214-212
- Antigen Symbol:
- Glutamate Receptor 1 (AMPA subtype)
- Antigen Name:
- AMPA 1
- Antigen Synonyms:
- GLUH1|AMPA-selective glutamate receptor 1|GluR-A|GluR-1|GluA1|GLUR 1|GluR K1|GLUR A|AMPA selective glutamate receptor 1
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC0657
- Reactivity:
- Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P42261
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 800-906 of human GluR1/GRIA1 (P42261)
- Sequence:
- ALSLSNVAGVFYILIGGLGLAMLVALIEFCYKSRSESKRMKGFCLIPQQSINEAIRTSTLPRNSGAGASSGGSGENGRVVSHDFPKSMQSIPCMSHSSGMPLGATGL
- Form:
- Liquid
- Molecular Weight:
- 102 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C