Anti-CARD8 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to CARD8 for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:200
Type: Primary
Antigen: CARD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76862-458
- Antigen Symbol:
- CARD8
- Antigen Name:
- Apoptotic protein NDPP 1
- Antigen Synonyms:
- CARDINAL|CARD8_HUMAN|CARD-inhibitor of NF-kappa-B-activating ligand|CARD 8|CARD inhibitor of NF kappaB activating ligands|CARD inhibitor of NF kappa B activating ligand|CARD inhibitor of NF kappaB activating ligand|Apoptotic protein NDPP1|CARD8
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9Y2G2
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CARD8 (NP_003796.1)
- Sequence:
- MEARDKQVLRSLRLELGAEVLVEGLVLQYLYQEGILTENHIQEINAQTTGLRKTMLLLDILPSRGPKAFDTFLDSLQEFPWVREKLKKAREEAMTDLPAGDRLTGIPSHILNSSPSDRQINQLAQRLGPEWEPMVLSLGLSQTDIYRCKANHPHNVQSQVVEAFIRWRQRFGKQATFQSLHNGLRAVEVDPSLLLHMLE
- Form:
- Liquid
- Molecular Weight:
- 60 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
Specifications
Frequently Bought Together