About this item
Rabbit polyclonal antibody to FGD4 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:200
Type: Primary
Antigen: FGD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76866-114
- Antigen Symbol:
- FGD4
- Antigen Name:
- 9030023J02Rik
- Antigen Synonyms:
- FGD4|FGD1 related F actin binding protein|CMT4H|FGD1-related F-actin-binding protein|DKFZp313E1818|9330209B17Rik|DKFZp434K1572|Actin filament binding protein frabin|Actin filament-binding protein frabin
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q96M96
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 99-198 of human FGD4 (NP_640334.2)
- Sequence:
- QMECEEEKAATLSSDTSIQASEPLLDTHIVNGERDETATAPASPTTDSCDGNASDSSYRTPGIGPVLPLEERGAETETKVQERENGESPLELEQLDQHHE
- Form:
- Liquid
- Molecular Weight:
- 100 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C