To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:80K-H protein
- Antigen Symbol:Glucosidase 2 subunit beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P14314
- Antigen Synonyms:AGE-R2|GLU2B_HUMAN|AGE-binding receptor 2|Glucosidase 2 subunit beta|Hepatocystin|Glucosidase II beta subunit|G19P1|Glucosidase II subunit beta|PCLD
- Storage Buffer:Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Molecular Weight:80 kDa
- Sequence:MAEVTREGFRLKKILIEDWKKAREEKQKKLIELQAGKKSLEDQVEMLRTVKEEAEKPEREAKEQHQKLWEEQLAAAKAQQEQELAADAFKELDDDMDGTVSVTELQTHPELDTDGDGALSEAEAQALLSGDTQTDATSFYDRVWAA
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 130-275 of human PRKCSH (P14314)
- Purification:Affinity purification.
- Cat. No.:76858-238
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Glucosidase 2 subunit beta for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:100
Type: Primary
Antigen: Glucosidase 2 subunit beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat