About this item
Rabbit monoclonal [ARC0509] antibody to Lactate Dehydrogenase for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Lactate Dehydrogenase
Clonality: Monoclonal
Clone: ARC0509
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77200-968
- Antigen Symbol:
- Lactate Dehydrogenase
- Antigen Name:
- Cell proliferation inducing gene 19 protein
- Antigen Synonyms:
- Epididymis secretory sperm binding protein Li 133P|HEL S 281|L lactate dehydrogenase B chain|L-lactate dehydrogenase A chain|Cell proliferation-inducing gene 19 protein|L lactate dehydrogenase A chain|Epididymis secretory protein Li 281|HEL S 133P|GSD11
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC0509
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# P00338
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant protein of human LDHA.
- Form:
- Liquid
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C