To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ALS10
- Antigen Symbol:TDP43
- Clonality:Monoclonal
- Clone:TARDP/349
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q13148
- Antigen Synonyms:OTTHUMP00000002173|TADBP_HUMAN|TDP 43|TAR DNA binding protein 43|OTTHUMP00000002171|OTTHUMP00000002172|TAR DNA-binding protein 43|TAR DNA binding protein|TARDBP
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:44 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment within human TDP43 aa 200 to the C-terminus
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77178-342
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [TARDP/349] antibody to TDP43 for ELISA, WB and IHC-P with samples derived from Human.
- Validated Applications: ELISA, WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: TDP43
Clonality: Monoclonal
Clone: TARDP/349
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human