To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Form:Liquid
- Gene ID:UniprotID# P04271
- Antigen Synonyms:S-100 protein beta chain|S100 calcium binding protein beta (neural)|S 100 calcium binding protein beta chain|S 100 protein beta chain|Protein S100 B|S100|S-100 protein subunit beta|Protein S100-B|S100 beta
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:10 - 12 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-92) of human S100B protein. The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Cat. No.:77178-040
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [S100B/4148] antibody to S100 beta for Flow Cytometry, IF and WB with samples derived from Human.
- Validated Applications: Flow Cytometry, IF, WB
- Recommended Dilutions: Flow Cytometry: 0.5 to 1 µg/million cells, IF: 1 to 2 µg/ml, WB: 0.5 to 1 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: S100 beta
Clonality: Monoclonal
Clone: S100B/4148
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human