To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:FLJ10128
- Antigen Symbol:UACA
- Clonality:Monoclonal
- Clone:UACA/1222
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9ZF9
- Antigen Synonyms:MGC141969|UACA_HUMAN|Uveal autoantigen with coiled coil domains and ankyrin repeats|NUCLING|KIAA1561|MGC141967|UACA|Nuclear membrane binding protein|Uveal autoantigen with coiled-coil domains and ankyrin repeats
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:160 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant human UACA protein
- Purification:Protein A/G chromatography
- Cat. No.:76967-924
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [UACA/1222] antibody to UACA for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1 to 2 µg/ml
Type: Primary
Antigen: UACA
Clonality: Monoclonal
Clone: UACA/1222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse