To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Cell proliferation inducing gene 46 protein
- Antigen Symbol:Cytokeratin 18
- Clonality:Monoclonal
- Clone:KRT18/836
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Dog,Cat
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P05783
- Antigen Synonyms:CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:45 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human KRT18 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76958-536
- Packaging:Plastic vial
Mouse monoclonal [KRT18/836] antibody to Cytokeratin 18 for Flow Cytometry, IF and IHC-P with samples derived from Human, Canine and Feline.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 18
Clonality: Monoclonal
Clone: KRT18/836
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Dog;Cat