Order Entry
United States
ContactUsLinkComponent
Anti-Interferon gamma + IL4 Rabbit Polyclonal Antibody
Anti-Interferon gamma + IL4 Rabbit Polyclonal Antibody
  77071-284
undefined
Anti-Interferon gamma + IL4 Rabbit Polyclonal Antibody
  77071-284
 :  A101413-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Host:
  • Isotype:
  • Reactivity:
  • Size:
    100 µg
  • Environmentally Preferable:
  • Cat. No.:
    77071-284

 

 

Rabbit polyclonal antibody to IL-4 for WB and ELISA with samples derived from Human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Interferon gamma + IL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human