About this item
Rabbit monoclonal [ARC57717] antibody to Pepsinogen II / PGC for IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC
- Recommended Dilutions: IHC: 1:50 to 1:200
Type: Primary
Antigen: Pepsinogen II / PGC
Clonality: Monoclonal
Clone: ARC57717
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.