To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ABC 10
- Antigen Symbol:ABCA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:24
- Antigen Synonyms:ABC A4|ABC transporter, retinal-specific|ABCA 4|ABCR|ARMD 2|ABCA4_HUMAN|ABC10|ARMD2|ABCA4
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.3, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-KQQTESHDLPLHPR
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the C terminus of human ABCA4.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:76961-070
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to ABCA4 for ELISA and IF with samples derived from Human.
- Validated Applications: ELISA, IF
Type: Primary
Antigen: ABCA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human