To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:3830405G04Rik
- Antigen Symbol:ABRA1
- Clonality:Monoclonal
- Clone:ABRA1-01
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q6UWZ7
- Antigen Synonyms:CCDC98|ABRA1|Abraxas protein|5630400M01Rik|AI506069|AV118690|BRCA1-A complex subunit Abraxas|AL024423|ABRAXAS
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein corresponding to amino acids 1-313 of ABRA1 with N-terminal His6 tag
- Purification:Protein A chromatography
- Cat. No.:76967-430
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [ABRA1-01] antibody to ABRA1 for Flow Cytometry, IP and WB with samples derived from Human.
- Validated Applications: Flow Cytometry, IP, WB
- Recommended Dilutions: Flow Cytometry: 1-2 µg/ml
Type: Primary
Antigen: ABRA1
Clonality: Monoclonal
Clone: ABRA1-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human