To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD158 antigen like family member D
- Antigen Symbol:KIR2DL4
- Clonality:Monoclonal
- Clone:mAb#33
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 T
- Environmentally Preferable:
- Form:Liquid
- Antigen Synonyms:CD158d|KI2L4_HUMAN|CD158 antigen-like family member D|CD_antigen=CD158d|Killer cell inhibitory receptor 103AS|Killer cell immunoglobulin like receptor 2DL4|Killer cell immunoglobulin-like receptor 2DL4|Killer cell immunoglobulin like receptor, two domains, long cytoplasmic tail, 4|G9P
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:NK3.3 cells and KIR2DL4-Ig fusion protein
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76965-188
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [mAb#33] antibody to KIR2DL4 (PE) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: KIR2DL4
Clonality: Monoclonal
Clone: mAb#33
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human