To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:LAMB2
- Antigen Symbol:Laminin gamma 1
- Clonality:Monoclonal
- Clone:A5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P11047, Q5VYE7, Q6NVY8
- Antigen Synonyms:Laminin B2 chain|Laminin-11 subunit gamma|Laminin-1 subunit gamma|Laminin, gamma 1 (formerly LAMB2)|Laminin subunit gamma-1|Laminin-10 subunit gamma|lamc1|LAMC1_HUMAN|Laminin gamma 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:210 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Murine EHS laminin preparation
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76965-942
- Packaging:Plastic vial
Specifications
About this item
Rat monoclonal [A5] antibody to Laminin gamma 1 for Flow Cytometry, IF and IHC-P with samples derived from Human and Mouse.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Laminin gamma 1
Clonality: Monoclonal
Clone: A5
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse