Order Entry
United States
ContactUsLinkComponent
Anti-GST Goat Polyclonal Antibody
Anti-GST Goat Polyclonal Antibody
Supplier:  ANTIBODIES.COM LLC
Certificates
undefined
Anti-GST Goat Polyclonal Antibody
Supplier:  ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 76835-448
  • 76834-084
  • Antigen Symbol:
  • GST
  • GST
  • Antigen Name:
  • Glutathione S Transferase
  • Glutathione S Transferase
  • Antigen Synonyms:
  • Glutathione S-transferase class-mu 26 kDa isozyme|GST epsilon|GST class alpha 2|GST|GST class alpha 1|GST gamma|Glutathione S transferase A1|Glutathione S transferase A2|GSTA1
  • Glutathione S-transferase class-mu 26 kDa isozyme|GST epsilon|GST class alpha 2|GST|GST class alpha 1|GST gamma|Glutathione S transferase A1|Glutathione S transferase A2|GSTA1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Schistosoma japonicum
  • Schistosoma japonicum
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Glutathione-S-Transferase, expressed in and purified from E. coli
  • Recombinant Glutathione-S-Transferase, expressed in and purified from E. coli
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Affinity purification
  • Affinity purification
  • Concentration:
  • 3 mg/ml
  • 3 mg/ml
  • Storage Buffer:
  • Supplied in Phosphate Buffered Saline with 20% Glycerol and 0.05% Sodium Azide
  • Supplied in Phosphate Buffered Saline with 20% Glycerol and 0.05% Sodium Azide
  • Storage Temperature:
  • Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
  • Shipping Temperature:
  • Shipped on blue ice at +4 °C.
  • Shipped on blue ice at +4 °C.

Specifications

Frequently Bought Together