To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TDH Toxin + TRH Toxin
- Clonality:Monoclonal
- Clone:vp-01
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P19249,Q769J9
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Culture supernatant of V. parahemolyticus
- Purification:Affinity purification
- Cat. No.:77072-746
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal (vp-01) antibody to TDH Toxin + TRH Toxin for WB and ELISA.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1,000
- Host: Mouse
- Clonality: Monoclonal
- Isotype: IgG1
- Conjugate: Unconjugated
Type: Primary
Antigen: TDH Toxin + TRH Toxin
Clonality: Monoclonal
Clone: vp-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: