Mouse monoclonal (ABT-TCL1) antibody to TCL1 for IHC with samples derived from Human.
- Validated Applications: IHC
Type: Primary
Antigen: Tcl1
Clonality: Monoclonal
Clone: ABT-TCL1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77063-968
- 77081-294
- Antigen Symbol:
- Tcl1
- Tcl1
- Antigen Name:
- Anti TCL1A
- Anti TCL1A
- Antigen Synonyms:
- P14 TCL1|Protein p14 TCL1|Lymphoma/leukemia, T-cell|P14 TCL1 protein|Oncogene TCL1|T cell lymphoma 1|T cell leukemia 1|Oncogene TCL-1|T cell lymphoma 1A
- P14 TCL1|Protein p14 TCL1|Lymphoma/leukemia, T-cell|P14 TCL1 protein|Oncogene TCL1|T cell lymphoma 1|T cell leukemia 1|Oncogene TCL-1|T cell lymphoma 1A
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-TCL1
- ABT-TCL1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P56279
- UniprotID# P56279
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human TCL1
- Synthetic peptide derived from human TCL1
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage Buffer:
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Shipping Temperature:
- Shipped on blue ice at +4 °C.
- Shipped on blue ice at +4 °C.
- Tested Applications:
- IHC
- IHC