To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:21 kDa pRb associated protein
- Antigen Symbol:EID1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9Y6B2
- Antigen Synonyms:Chromosome 15 open reading frame 3|CBP|C15orf3|CREBBP/EP300 inhibitory protein 1|21 kDa pRb-associated protein|E1A like inhibitor of differentiation|CRI 1|CREBBP/EP300 inhibitor 1|CRI1
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:20 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human EID1 (amino acids 71-120)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77076-950
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to EID1 for WB, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: EID1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human