Anti-NOM1 Rabbit Polyclonal Antibody
100 µg, Plastic vial
Catalog # 77078-958
Supplier: Antibodies.com
Supplier #: A99207-100
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit polyclonal antibody to NOM1 for IHC and ELISA with samples derived from Human and Mouse.
- Recommended dilutions: IHC: 1:50 to 1:100 ELISA: 1:40000
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: NOM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Documents
Specifications
- Size:100 µg
- Packaging:Plastic vial
- Clonality:Polyclonal
- Molecular Weight:96 kDa
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Form:Liquid
- Storage Buffer:Supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% sodium azide, and 50% glycerol
- Tested Applications:IHC
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77078-958
- Reactivity:Human|Mouse
- Shipping Temperature:Shipped on blue ice at 4 °C
- Antibody Type:Primary
- Antigen Symbol:NOM1
- Gene ID:UniprotID# Q5C9Z4
- Immunogen:Synthetic peptide derived from human NOM1 (amino acids 661-710)
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:C7orf3|Nucleolar MIF4G domain-containing protein 1|SGD1 homolog
- Antigen Name:C7orf3
- ELISA:Yes
- Storage Temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.



