Rabbit polyclonal antibody to RAPGEF6 for WB with samples derived from Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: RAPGEF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77216-494
- Antigen Symbol:
- RAPGEF6
- Antigen Name:
- DKFZp667N084
- Antigen Synonyms:
- PDZ domain-containing guanine nucleotide exchange factor 2|PDZ GEF 2|PDZ domain containing guanine nucleotide exchange factor 2|PDZ-GEF2|PDZ domain containing guanine nucleotide exchange factor (GEF) 2|PDZ domain containing guanine nucleotide exchange factor I|KIA001LB|DKFZp686I15116|PDZ GEF2
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q8TEU7
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 145-240 of human RAPGEF6 (NP_001157858)
- Sequence:
- INYKGERQTITDDVEVNSYLSLPADLTKMHLTENPHPQVTHVSSSQSGCSIASDSGSSSLSDIYQATESEVGDVDLTRLPEGPVDSEDDEEEDEEI
- Form:
- Liquid
- Molecular Weight:
- 179 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C