DNA/RNA Shield Fecal Collection Tube (with Beads)
Catalog # 77020-304
Supplier: Zymo Research
CAS Number:
Specifications
- Application:
- Method / Format:Bead beating
- Prep size:Scalable
- Refine target molecule:Bacterial DNA
- Sample type:Stool
- Target molecule:Copurification of DNA & RNA
- Description:DNA/RNA shield fecal collection tube (with beads)
- Size:10 pack
- Environmentally Preferable:
- Cat. No.:77020-304
Specifications
About this item
DNA/RNA shield fecal collection tubes are designed for the collection and preservation of nucleic acids from stool specimens.
- Pre-filled with DNA/RNA shield to inactivate pathogens and stabilize microbiome profiles (DNA and RNA) at ambient temperature
DNA/RNA Shield Fecal Collection tubes are designed for the collection and preservation of nucleic acids from stool specimens. These fecal collection tubes take a microbial snapshot of a sample while inactivating viruses making samples safe and ready for transport. Samples stored in these fecal collection tubes are stable at ambient temperature, and can be frozen for longer-term storage. Simply collect a spoonful of the specimen into the stool collection tubes prefilled with preservative and shake vigorously to ensure proper stabilization. R1137 is pre-filled with 5 glass beads to help facilitate homogenization.