Order Entry
United States
ContactUsLinkComponent
Quick-16S Plus NGS Library Prep Kit (V4)
Quick-16S Plus NGS Library Prep Kit (V4)
Supplier:  Zymo Research
Certificates
Quick-16S Plus NGS Library Prep Kit (V4)
Supplier:  Zymo Research

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Environmentally Preferable:

Specifications

Frequently Bought Together