Anti-MOCS3 Rabbit Polyclonal Antibody
Catalog # 76757-604
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Adenylyltransferase and sulfurtransferase MOCS3 {ECO:0000255|HAMAP-Rule:MF_03049}
- Antigen Symbol:MOCS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:27304
- Antigen Synonyms:Adenylyltransferase MOCS3 {ECO:0000255|HAMAP-Rule:MF_03049}|Molybdopterin synthase sulfurylase|27780 {ECO:0000255|HAMAP-Rule:MF_03049}|Molybdenum cofactor synthesis protein 3 {ECO:0000255|HAMAP-Rule:MF_03049}|Sulfurtransferase MOCS3 {ECO:0000255|HAMAP-Rule:MF_03049}|Molybdopterin-synthase adenylyltransferase {ECO:0000255|HAMAP-Rule:MF_03049}|Sulfur carrier protein MOCS2A adenylyltransferase {ECO:0000255|HAMAP-Rule:MF_03049}|Adenylyltransferase and sulfurtransferase MOCS3 {ECO:0000255|HAMAP-Rule:MF_03049}|Sulfur carrier protein MOCS2A sulfurtransferase {ECO:0000255|HAMAP-Rule:MF_03049}|MPT synthase sulfurylase|Molybdopterin-synthase sulfurtransferase {ECO:0000255|HAMAP-Rule:MF_03049}|MOCS3 {ECO:0000255|HAMAP-Rule:MF_03049}|28111 {ECO:0000255|HAMAP-Rule:MF_03049}
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 271-460 of human MOCS3 (NP_055299.1).
- Purification:Affinity purification
- Cat. No.:76757-604
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MOCS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat