Human Recombinant CD24 Protein (from HEK293 cells)
Catalog # 77132-852
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Environmentally Preferable:
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:100133941
- Protein Synonyms:CD24A|CD24
- Protein/Peptide Name:CD24
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:5.0 kDa
- Sequence:Ser 27 - Gly 59
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77132-852