Order Entry
United States
ContactUsLinkComponent
 
SARS-CoV-2 (COVID-19) Recombinant Nucleocapsid (N) Protein (from HEK293 cells)
  77132-590
 :  Prosci
 :  
SARS-CoV-2 (COVID-19) Recombinant Nucleocapsid (N) Protein (from HEK293 cells)
  77132-590
 :  Prosci
 :  10-080
 :  

 

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    SARS-CoV-2
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Storage Conditions:
    We recommended to store this recombinant protein as received at 2...8 °C for up to one month. For longer-term storage, aseptically aliquot in working volumes without diluting and store at −80 °C. Avoid repeated freeze thaw cycles.
  • Protein/Peptide Name:
    Nucleocapsid (N)
  • Purity:
    >95% by SDS PAGE
  • Molecular Weight:
    The predicted molecular weight of Recombinant SARS-CoV-2, Nucleocapsid (N) Protein is Mr 47 kDa
  • Sequence:
    MSDNGPQNQRNAPRITFGGPSDSTGSNQNGERSGARSKQRRPQGLPNNTASWFTALT
    QHGKEDLKFPRGQGVPINTNSSPDDQIGYYRRATRRIRGGDGKMKDLSPRWYFYYLGTG
    PEAGLPYGANKDGIIWVATEGALNTPKDHIGTRNPANNAAIVLQLPQGTTLPKGFYAEGS
    RGGSQASSRSSSRSRNSSRNSTPGSSRGTSPARMAGNGGDAALALLLLDRLNQLESKMS
    GKGQQQQGQTVTKKSAAEASKKPRQKRTATKAYNVTQAFGRRGPEQTQGNFGDQELIR
    QGTDYKHWPQIAQFAPSASAFFGMSRIGMEVTPSGTWLTYTGAIKLDDKDPNFKDQVILL
    NKHIDAYKTFPPTEPKKDKKKKADETQALPQRQKKQQTVTLLPAADLDDFSKQLQQSMSS
    ADSTQA
  • Concentration:
    0.5 mg/ml
  • Formulation:
    Liquid. This recombinant protein was 0.2 µm filtered and Lyophilized from modified Dulbecco’s phosphate buffered saline (1X PBS) pH 7.2 - 7.3 with no calcium, magnesium, or preservatives.
  • Shipping Temperature:
    Dry Ice
  • Tested Applications:
    ELISA,WB
  • Cat. No.:
    77132-590