Anti-PRDM9 Recombinant Antibody [Clone: RAB-C370]
Catalog # 76630-840
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:PR/SET domain 9
- Antigen Symbol:PRDM9
- Clonality:Recombinant
- Clone:RAB-C370
- Conjugation:Unconjugated
- ELISA:Yes
- Host:
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:Fab fragment kappa
- Reactivity:Human
- Size:0.1 mg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Gene ID:56979
- Antigen Synonyms:PR domain zinc finger protein 9|PR domain-containing protein 9|PRDM9-A003-1459|[histone H4]-N-methyl-L-lysine20 N-methyltransferase PRDM9|EC:2.1.1.-|[histone H3]-lysine36 N-trimethyltransferase PRDM9|[histone H4]-lysine20 N-methyltransferase PRDM9|[histone H3]-lysine4 N-trimethyltransferase PRDM9|Histone-lysine N-methyltransferase PRDM9|Protein-lysine N-methyltransferase PRDM9|[histone H3]-lysine9 N-trimethyltransferase PRDM9|anti-PRDM9-RAB-C370
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM9 protein under non-denaturing conditions.
- Cat. No.:76630-840
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PR/SET domain 9
Clonality: Recombinant
Clone: RAB-C370
Conjugation: Unconjugated
Host: Recombinant
Isotype: Fab fragment kappa
Reactivity: Human