Order Entry
United States
ContactUsLinkComponent
Cell Counting Kit-8 (CCK-8)
Cell Counting Kit-8 (CCK-8)
Catalog # 76466-042
CAS Number:  
undefined
Cell Counting Kit-8 (CCK-8)
Catalog # 76466-042
Supplier Number:  AR1160
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    6 mg/ml
  • Description:
    Cell counting kit, liquid
  • Size:
    500 ml, 10 μl per well
  • Environmentally Preferable:
  • Cat. No.:
    76466-042
  • Sensitivity:
    CCK-8 is the highest sensitive dye for the cell based assay.
  • Buffer volume:
    9% NaCl
  • Storage temperature:
    −4 °C (protect from light)
  • No. of tests:
    500 assays

Specifications

About this item

Cell counting kit-8 (CCK-8) allows very convenient assays by utilizing highly water-soluble tetrazolium salt. WST-8 [2- (2- methoxy-4-nitrophenyl)-3- (4-nitrophenyl)-5- (2, 4-disulfophenyl)-2H- tetrazolium, monosodium salt]* produces a water-soluble formazan dye upon reduction in the presence of an electron carrier. Cell Counting Kit-8 is a one-bottle solution; no premixing of components is required.

  • One bottle, ready to use solution
  • No organic solvents or isotopes required
  • No harvesting, no washing and no solubilization steps
  • No toxicity to cell
  • More sensitive than MTT, XTT, MTS or WST-1

Cell Counting Kit-8, being nonradioactive, allows sensitive colorimetric assays for the determination of the number of viable cells in cell proliferation cytotoxicity assays. WST-8 is reduced by dehydrogenases in cells to give an orange colored product (formazan), which is soluble in the tissue culture medium. The amount of the formazan dye generated by the activity of dehydrogenases in cells is directly proportional to the number of living cells.

The cell proliferation assay using CCK-8 correlates well with the [(3)H] thymidine incorporation assay. And the CCK-8 assay can also be substituted for the [(3)H]thymidine incorporation assay. The detection sensitivity of CCK-8 is higher than other tetrazolium salts such as MTT, XTT, MTS or WST-1.