ContactUsLinkComponent
VWR® Premium Laboratory Refrigerators
VWR® Premium Laboratory Refrigerators
Supplier:  VWR International
   Sustainable Options Available
Certificates
undefined
VWR® Premium Laboratory Refrigerators
Supplier:  VWR International
   Sustainable Options Available
Specifications
  • Catalog No:
  • 76919-388
  • 76919-378
  • 76919-380
  • 76919-386
  • Environmentally Preferable:
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Cat. No.:
  • 76919-388
  • 76919-378
  • 76919-380
  • 76919-386
  • Defrost:
  • Cycle defrost
  • Cycle defrost
  • Cycle defrost
  • Cycle defrost
  • Shelves/drawers:
  • 10 adjustable shelves
  • 5 adjustable shelves
  • 10 adjustable shelves
  • 5 adjustable shelves
  • Door Type:
  • 2 solid swing doors
  • 1 glass swing door, right hinged
  • 2 glass swing doors
  • 1 solid swing door, right hinged
  • Weight:
  • 223 kg (491 lbs.)
  • 159 kg (350 lbs.)
  • 264 kg (581 lbs.)
  • 124 kg (274 lbs.)