The all-in-one imager for any imaging application. From gel analysis and documentation to in vivo fluorescence and bioluminescence animal imaging, the UVP Biospectrum Advanced does it all.
- Two high performance cooled camera options
- Illumination sources - overhead laser bars in RGBW, NIR, and violet (405 nm)
- Motorized lift for sample magnification range
- Included warming plate or UV transilluminator
- VisionWorks software for one-click workflows
The UVP BioSpectrum Advanced is an all in-one BioImaging system with in vivo imaging capabilities for sample documentation and analyzes, including in vivo animal imaging, plant imaging, chemiluminescence, NIR, and fluorescence. This all-in-one system is perfect for any lab within research institutions, medical facilities, academic research institutions, pharmaceutical companies and more.
The UVP BioSpectrum Advanced comes with a choice of deeply-cooled CCD camera sensors capable of imaging fluorescence, chemiluminescence, and bioluminescence. The system features extended NIR, violet (405 nm), red, green, and blue laser bars, white epi (overhead) illumination, motorized platform lift and filter wheel, and automated lens. Add a sample warming plate, UV light transilluminator, or blue light transilluminator to fit your specific application needs.
- Catalog No:
- 76837-120
- 76837-096
- 76837-116
- 76837-114
- 76837-094
- 76837-108
- W×D×H:
- 24.5×18.2×36.0"
- 24.5×18.2×36.0"
- 24.5×18.2×36.0"
- 24.5×18.2×36.0"
- 24.5×18.2×36.0"
- 24.5×18.2×36.0"