Order Entry
United States
ContactUsLinkComponent
Anti-C9orf153 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-C9orf153 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  76107-830
 :  Bioss
Anti-C9orf153 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  76107-830
 :  Bioss
 :  BS-9498R-A750
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    C9orf153
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µl
  • Environmentally Preferable:
  • Epitope:
    1-80/101
  • Form:
    Liquid
  • Gene ID:
    389766
  • Antigen Synonyms:
    MGC131702|Chromosome 9 open reading frame 153|bA507D14.1|Uncharacterized protein C9orf153|Hypothetical protein LOC389766
  • Modification:
    Unmodified
  • Storage Buffer:
    Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
  • Storage Temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping Temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C9orf153
  • Purification:
    Purified by Protein A
  • Cat. No.:
    76107-830
  • Packaging:
    Vial

 

 

C9orf153 is a 101 amino acid protein that exists as two alternatively spliced isoforms. The gene encoding C9orf153 maps to human chromosome 9q21.33. Chromosome 9 consists of about 145 million bases, represents 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses the largest interferon family gene cluster. Chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias.

Type: Primary
Antigen: C9orf153
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-80/101
Host: Rabbit
Isotype: IgG
Reactivity: Human