To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:KChIP2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Epitope:1-100/270
- Form:Liquid
- Gene ID:30819
- Antigen Synonyms:A-type potassium channel modulatory protein 2|Kv channel-interacting protein 2|DKFZp566L1246|KCIP2_HUMAN|KChIP 2|MGC17241|A type potassium channel modulatory protein 2|KChIP2|Cardiac voltage gated potassium channel modulatory subunit|Potassium channel-interacting protein 2|Kcnip2|KCNIP 2|Kv channel interacting protein 2|Potassium channel interacting protein 2|KCNIP-2|Cardiac voltage-gated potassium channel modulatory subunit
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human KChIP2
- Purification:Purified by Protein A
- Cat. No.:76109-296
- Packaging:Vial
Specifications
About this item
The downstream regulatory element, DRE, acts as a location-dependent gene silencer. DREAM (for DRE-antagonist modulator) is a Ca²⁺ regulated transcriptional repressor that specifically binds to the DRE. DREAM regulates transcription of prodynorphin and c-Fos genes and shows 99% nucleotide homology to the Kv channel-interacting proteins (KChIPs). KChIP family members include KChIP1, which is expressed in brain, KChIP2, which is expressed in heart, brain, and lung, KChIP3 (also designated calsenilin), which is expressed in brain and testis and KChIP4.
Type: Primary
Antigen: KChIP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/270
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat