To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:ECHS1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Epitope:111-210/290
- Form:Liquid
- Gene ID:54981
- Antigen Synonyms:Enoyl CoA hydratase short chain 1 mitochondrial|Enoyl Coenzyme A hydratase short chain 1 mitochondrial|C80529|Enoyl Coenzyme A hydratase short chain 1|EC 4.2.1.17|Enoyl CoA hydratase 1|Short chain enoyl CoA hydratase|Mitochondrial short chain enoyl coenzyme A hydratase 1|Enoyl CoA hydratase mitochondrial|OTTHUMP00000020811|SCEH|ECHS 1
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:31 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ECHS1
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76079-638
- Packaging:Vial
ECHS1 (Enoyl Coenzyme A hydratase, short chain, 1) catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs, in the second step of the mitochondrial fatty acid beta-oxidation pathway.
Type: Primary
Antigen: ECHS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 111-210/290
Host: Rabbit
Isotype: IgG
Reactivity: