Anti-HIF Prolyl Hydroxylases Rabbit Polyclonal Antibody (FITC)
Catalog # 10319-850
Supplier: Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:HIF Prolyl Hydroxylases
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Epitope:201-300/502
- Form:Liquid
- Antigen Synonyms:PH 4|EGLN4|Proline 4 hydroxylase|Prolyl 4 hydroxylase transmembrane (endoplasmic reticulum)|Prolyl hydroxlase domain containing 4|PH4|P4htm|HIF-prolyl hydroxylase 4|HIFPH4|Transmembrane prolyl 4-hydroxylase|Hypoxia-inducible factor prolyl hydroxylase 4|PHD4|P4H with transmembrane domain|FLJ20262|P4HTM_HUMAN|HPH-4|Hypoxia inducible factor prolyl 4 hydroxylase|HIF-PH4|P4H TM|HIF prolyl hydroxylase PH4|P4H-TM|Hypoxia inducible factor prolyl hydroxylase
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:57 kDa
- Storage Temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HIF Prolyl Hydroxylases
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:10319-850
- Packaging:Vial
Specifications
About this item
HIF Prolyl Hydroxylases.
Type: Primary
Antigen: HIF Prolyl Hydroxylases
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 201-300/502
Host: Rabbit
Isotype: IgG
Reactivity: Mouse