Deflagrating Spoon and Cap
Catalog # 470346-122
Supplier: Wards
Specifications
- Description:Deflagrating spoon and cap, 12/pack
- Environmentally Preferable:
- Cat. No.:470346-122
Specifications
About this item
Set of 12 deflagrating spoons and caps, featuring a steel cup and rod with an aluminum cap, designed for controlled combustion reactions and experiments in laboratory settings.
- Package of 12 metal deflagration spoons and caps - Spoons measure approximately 35 cm in length, and caps measure approximately 90 mm in diameter
- Spoon bowl measures approximately 15 mm in diameter
- Long spoon and wide cap protect user from extreme exothermic reactions
- Pack of twelve is the perfect size for supplying chemistry labs
- Spoons are great in both professional and educational lab settings
This bundle contains enough spoons to supply a small lab in a school or in a research setting. These spoons are often used for burning sulfur, phosphorus, and other heavily reactive materials. Deflagrating spoons are perfect for handling strong exothermic reactions such as these, allowing the user to move the material while keeping a safe distance. The cap attaches to the spoon and provides another layer of safety.