About this item
VIC phosphoramidite is the most convenient building block to incorporate VIC tag into an oligonucleotide.
- Emits in the green-yellow part of the visible spectrum
- Multiple applications
VIC is a fluorescent dye that was originally developed by Applied Biosystems (now, Thermofisher). It is used to fluorescently label oligonucleotides at the 5’-end (for use as probes in a variety of real-time PCR, hybridization and fluorescence-based genetic analysis applications).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Environmentally Preferable: