Order Entry
United States
ContactUsLinkComponent
2D Coded Sample Storage Tubes, 0.2 ml, 240-format, External Thread, Azenta Life Sciences
2D Coded Sample Storage Tubes, 0.2 ml, 240-format, External Thread, Azenta Life Sciences
Supplier:  AZENTA US, INC
Certificates
undefined
2D Coded Sample Storage Tubes, 0.2 ml, 240-format, External Thread, Azenta Life Sciences
Supplier:  AZENTA US, INC
Certificates

Specifications

  • Catalog No:
  • 76636-616
  • 76637-006
  • 76636-618
  • 76636-832
  • Height with cap:
  • 27.5 mm
  • 27.5 mm
  • 27.5 mm
  • 27.5 mm
  • Ext.Ø with cap:
  • 5.8 mm
  • 5.8 mm
  • 5.8 mm
  • 5.8 mm
  • Centre to centre:
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • Height in rack:
  • 28.0 mm
  • 28.0 mm
  • 28.0 mm
  • 28.0 mm
  • Overall rack height incl. lid:
  • 31.0 mm
  • 31.0 mm
  • 31.0 mm
  • 31.0 mm

Specifications

Empty racks

Product Information