PCR Clear Adhesive Seals, Peelable
Supplier: AZENTA US, INC
Certificates
About this item
The adhesive PCR seal is a durable transparent polyester film with a strong adhesive layer. Recommended for PCR, qPCR, and other optical applications.
- Non-pierceable
- Peelable
- Allows for sealing of complete 96 well plates, but also individual or multiple Breakable Horizontally or Vertically or breakable vertically
- Adhesive-free end tabs for easy removal
- No sticky residue on the plate surface following removal
- Temperature range: –20 to +110 °C
- Non sterile
Adhesive seals provide a versatile, yet robust, method for protecting samples.
To obtain the best sealing results with adhesive seals, we strongly recommend the use of high quality plates with raised rim sealing rings for optimal sealing integrity and guaranteed flatness. All Azenta PCR plates are designed with these features.
Accessories information: Adhesive seal roller; ensures consistent seal application across all wells, 1 roller.
Frequently Bought Together