To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Neuron specific enolase
- Clonality:Monoclonal
- Clone:VI-H14
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:6 ml
- Environmentally Preferable:
- Cat. No.:76634-502
This antibody is specific to γ-γ enolase isozyme of 46 kDa. This antibody stains neurons, neuroendocrine cells, gangliomas, oat cell carcinomas, astrocytomas, paragangliomas, glioblastomas, pheochromocytomas, melanomas and schwannomas.