Villin is One of the gelsolin family of calcium regulated actin binding proteins. Normally expressed in the brush border of epithelial cells lining the gastrointestinal tract, hepatobiliary tract and renal proximal convoluted tubules. In colorectal carcinoma, villin is highly expressed. In practice, villin can be included in the panel used for metastatic carcinoma to detect colorectal origin.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76634-226
- Antigen Name:
- Villin
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 1D2C3
- Reactivity:
- Human
- Host:
- Mouse
- Isotype:
- IgG1 kappa