To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Renal cell carcinoma
- Clonality:Monoclonal
- Clone:PN-15
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:6 ml
- Environmentally Preferable:
- Cat. No.:76634-440
Specifications
About this item
This antibody recognizes a 200 kDa glycoprotein (gp200) localized in the brush border of the proximal renal tubules and the luminal surface of Bowman’s capsule. gp200 was expressed by 93% of primary and 84% of metastatic renal carcinomas. Gp200 glycoprotein is also localized in the breast lobules and ducts, parathyroid glands, and epididymis.