Bacto™ TC Lactalbumin Hydrolysate, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Bacto TC Lactalbumin Hydrolysate, an animal origin peptone, is the enzymatically hydrolyzed protein portion of milk Whey.
- Animal origin (AO) peptone
- Mixture of peptides, amino acids, and carbohydrates (simple and complex)
- Can be used in mammalian, bacterial and insect cultures
- Used in recombinant proteins applications
- Used in animal health vaccines
Bacto TC Lactalbumin Hydrolysate, an animal origin peptone, is the enzymatically hydrolyzed protein portion of milk whey. This supplement is a mixture of peptides, amino acids, and carbohydrates, both simple and complex. Customers have successfully used Bacto TC Lactalbumin Hydrolysate to support the growth of many mammalian, bacterial, and insect cell types, and it has been used to produce both recombinant proteins and animal health vaccines. Customers have also used this supplement as an amino acid supplement for mammalian cell culture media.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together