PCR Plates, 96-Well, Vertically and Horizontally Breakable, Non-Skirted, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
96 well non-skirted plate, vertically and horizontally perforated, tears easily into strips or part plates.
- The correct number of wells can be separated off for each experiment, saving time and costs
- Available perforated either vertically, tearing into 8-well strips (⁹⁶/₈), or horizontally, tearing into 12-well strips (⁹⁶/₁₂)
- The new 12-strip 96/12 plates allow you to make full use of your gradient PCR instruments
- Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
- Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
- Standard profile: <0.2 ml (200 µl) working volume, 0.3 ml (300 µl) total well capacity when used with sealing options
- Alphanumeric grid reference to aid well and sample identification
- Compatible with standard multichannel pipettes
Based on the 96 Well non-skirted PCR plate, plates can quickly and easily be divided along the perforations between the rows allowing for the most flexible, efficient and cost-effective use of a PCR. The correct number of wells can be separated off for each experiment, saving time and costs.
All of these PCR plates are molded from virgin polypropylene under ISO certified cleanroom conditions in a UK-based production facility, and as such comply with the same stringent requirements as the FrameStar® range.
Frequently Bought Together