Sample Storage Tube Caps, Screw Caps
Supplier: AZENTA US, INC
Certificates
About this item
Tube screw caps are manufactured from high-quality virgin polypropylene and are designed for optimal seal quality and sample security.
- High chemical resistance
- Broad operating temperature range –196 to +121 °C, do not use in liquid phase nitrogen
- Automation friendly, available in Azenta cap carrier for use with automated capping and de-capping systems
- Autoclavable
- Available in up to 10 different colors to aid sample identification
Developed to exceed the demands of sample security, management and tracking in modern high-density storage applications.
Compatible with all Azenta 96-format sample storage tubes with a screw top, caps are available for use with either external or internal thread Azenta screw top tubes.
Ordering information: These tubes are supplied in bags of 960 caps.