Pro-Wipe® AP All-Purpose Industrial Rayon/Polyester Non-woven Wipes, Berkshire
76424-328
- Description:Pro-Wipe® AP All-purpose industrial rayon/polyester non-woven wipe
- Material:Polyester 50%/rayon 50%
- Environmentally Preferable:
- Cat. No.:76424-328
- Packaging:Flat pack; 150 sheets/pack
- W×L:23×23 cm (9×9")
Pro-Wipe® AP are tough, low-linting non-woven wipes.
- Chemically compatible with common cleaning and disinfecting solutions
- Low-linting and abrasion-resistant
- Knife cut edge
- Autoclavable
- Absorbs water and oil based solutions
- Smooth and soft with high strength and durability
- Highly absorbent core due to inner layer of cellulose
Pro-Wipe®AP is an all-purpose industrial rayon/polyester nonwoven wipe. Durable and low-linting, Pro-Wipe ®AP is tough enough to be a rag replacement and clean enough to be used in a controlled manufacturing environment.