ContactUsLinkComponent
Evergreen Transport Vial Rack, Caplugs Evergreen
Evergreen Transport Vial Rack, Caplugs Evergreen
Catalog # 76521-502
Supplier:  Caplugs
undefined
Evergreen Transport Vial Rack, Caplugs Evergreen
Catalog # 76521-502
Supplier:  Caplugs
Supplier Number:  240-3300-G10
Specifications
  • Color:
    Dark green
  • Environmentally Preferable:
  • Cat. No.:
    76521-502
  • No. of places:
    50
  • W×D×H:
    147×307×15 mm
Specifications
About this item
Caplugs Evergreen Transport Vial Rack is designed to hold up to fifty transport vials of any size.
  • Made up of polystyrene